Assay Details
| Gene Symbol: Osbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152818.2 | NP_690031.2 | 1 - 2 | 663 | 63 | |
| GenBank mRNA | AK049816.1 | - | 1 - 2 | 664 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbp2
Genomic Map