Assay Details
| Gene Symbol: Osbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152818.2 | NP_690031.2 | 3 - 4 | 1137 | 67 | |
| GenBank mRNA | BC058356.1 | - | 2 - 3 | 280 | 5696357 | 67 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058356.1 transcripts.
Set Membership: |
|

Osbp2
Genomic Map