Assay Details
| Gene Symbol: Osbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152818.2 | NP_690031.2 | 8 - 9 | 1893 | 100 | |
| GenBank mRNA | AK007088.1 | - | 4 - 5 | 474 | 100 | |
| AK044598.1 | - | 5 - 6 | 682 | 100 | ||
| BC031794.1 | - | 6 - 7 | 856 | 4501119 | 100 | |
| BC051185.1 | - | 6 - 7 | 853 | 100 | ||
| BC058356.1 | - | 7 - 8 | 1036 | 5696357 | 100 | |
| BC058602.1 | - | 6 - 7 | 856 | 6467653 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbp2
Genomic Map