Assay Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029291.1 | NP_083567.1 | 3 - 4 | 288 | 61 | |
| GenBank mRNA | AK048876.1 | - | 3 - 4 | 322 | 61 | |
| AK051387.1 | - | 3 - 4 | 337 | 61 | ||
| AK148105.1 | - | 3 - 4 | 320 | 61 | ||
| BC013537.1 | - | 3 - 4 | 288 | 4240784 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ascc2
Genomic Map