Assay Details
| Gene Symbol: Ascc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029291.1 | NP_083567.1 | 9 - 10 | 956 | 136 | |
| GenBank mRNA | AK048876.1 | - | 9 - 10 | 990 | 136 | |
| AK051387.1 | - | 9 - 10 | 1005 | 136 | ||
| AK148105.1 | - | 9 - 10 | 988 | 136 | ||
| BC013537.1 | - | 9 - 10 | 956 | 4240784 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ascc2
Genomic Map