Assay Details
| Gene Symbol: Grwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153419.2 | NP_700468.2 | 1 - 2 | 201 | 73 | |
| GenBank mRNA | AB001539.1 | - | 2 - 3 | 73 | ||
| AK077611.1 | - | 1 - 2 | 207 | 73 | ||
| BC008121.1 | - | 1 - 2 | 178 | 73 | ||
| BC083143.1 | - | 1 - 2 | 189 | 73 | ||
| BQ886252.1 | - | 1 - 2 | 204 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grwd1
Genomic Map