Assay Details
| Gene Symbol: Grwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153419.2 | NP_700468.2 | 2 - 3 | 309 | 87 | |
| GenBank mRNA | AB001539.1 | - | 3 - 4 | 381 | 87 | |
| AK077611.1 | - | 2 - 3 | 315 | 87 | ||
| BC008121.1 | - | 2 - 3 | 286 | 87 | ||
| BC083143.1 | - | 2 - 3 | 297 | 87 | ||
| BQ886252.1 | - | 2 - 3 | 312 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grwd1
Genomic Map