Assay Details
| Gene Symbol: Grwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153419.2 | NP_700468.2 | 4 - 5 | 691 | 82 | |
| GenBank mRNA | AB001539.1 | - | 5 - 6 | 763 | 82 | |
| AK077611.1 | - | 4 - 5 | 697 | 82 | ||
| BC008121.1 | - | 4 - 5 | 668 | 82 | ||
| BC028896.1 | - | 3 - 4 | 560 | 3674738 | 82 | |
| BC083143.1 | - | 4 - 5 | 679 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grwd1
Genomic Map