Assay Details
| Gene Symbol: Sgsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172718.3 | NP_766306.2 | 22 - 23 | 3016 | 81 | |
| NR_027934.1 | 13 - 14 | 2203 | 81 | |||
| GenBank mRNA | AK053555.1 | - | 22 - 23 | 2950 | 81 | |
| AK167632.1 | - | 13 - 14 | 2167 | 81 | ||
| BC138049.1 | - | 22 - 23 | 2942 | 8861068 | 81 | |
| BC138050.1 | - | 22 - 23 | 2942 | 8861069 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm1
Genomic Map