Assay Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023799.3 | NP_076288.1 | 11 - 12 | 2228 | 154 | |
| GenBank mRNA | AF132214.1 | - | 11 - 12 | 2073 | 154 | |
| AK129188.1 | - | 11 - 12 | 2457 | 154 | ||
| BC054821.1 | - | 11 - 12 | 2228 | 6515250 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgea5
Genomic Map