Assay Details
| Gene Symbol: Mgea5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023799.3 | NP_076288.1 | 13 - 14 | 2416 | 109 | |
| GenBank mRNA | AF132214.1 | - | 13 - 14 | 2261 | 109 | |
| AK014781.1 | - | 1 - 2 | 42 | 109 | ||
| AK077613.1 | - | 1 - 2 | 938 | 109 | ||
| BC054821.1 | - | 13 - 14 | 2416 | 6515250 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgea5
Genomic Map