Assay Details
| Gene Symbol: Pprc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK122322.1 | - | 2 - 3 | 331 | 83 | |
| AK147947.1 | - | 2 - 3 | 354 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pprc1| Gene Symbol: Pprc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK122322.1 | - | 2 - 3 | 331 | 83 | |
| AK147947.1 | - | 2 - 3 | 354 | 83 | ||
Set Membership: |
|