Assay Details
| Gene Symbol: Pprc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081214.1 | NP_001074683.1 | 4 - 5 | 627 | 63 | |
| GenBank mRNA | AK122322.1 | - | 4 - 5 | 585 | 63 | |
| AK147947.1 | - | 4 - 5 | 608 | 63 | ||
| BC066048.1 | - | 4 - 5 | 581 | 6825158 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pprc1
Genomic Map