Assay Details
| Gene Symbol: Pprc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081214.1 | NP_001074683.1 | 8 - 9 | 3674 | 107 | |
| GenBank mRNA | AK122322.1 | - | 8 - 9 | 3635 | 107 | |
| AK147947.1 | - | 8 - 9 | 3655 | 107 | ||
| AK171995.1 | - | 3 - 4 | 2267 | 107 | ||
| BC013720.1 | - | 4 - 5 | 1041 | 107 | ||
| BC066048.1 | - | 8 - 9 | 3628 | 6825158 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pprc1
Genomic Map