Assay Details
| Gene Symbol: Fbxl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133694.2 | NP_598455.2 | 2 - 3 | 501 | 74 | |
| GenBank mRNA | AK003032.1 | - | 2 - 3 | 224 | 74 | |
| AK145009.1 | - | 2 - 3 | 461 | 75 | ||
| AK172339.1 | - | 3 - 4 | 398 | 74 | ||
| BC016499.1 | - | 2 - 3 | 476 | 4221584 | 74 | |
| CK791452.1 | - | 2 - 3 | 187 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbxl15
Genomic Map