Assay Details
| Gene Symbol: Rwdd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203507.3 | NP_987103.1 | 3 - 4 | 490 | 82 | |
| GenBank mRNA | AK004597.1 | - | 3 - 4 | 444 | 82 | |
| AK005797.1 | - | 3 - 4 | 475 | 82 | ||
| AK016505.1 | - | 3 - 4 | 464 | 82 | ||
| AK185756.1 | - | 2 - 3 | 143 | 82 | ||
| BC016198.1 | - | 3 - 4 | 426 | 4503404 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rwdd4a
Genomic Map