Assay Details
| Gene Symbol: Psmd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026000.2 | NP_080276.2 | 1 - 2 | 246 | 65 | |
| GenBank mRNA | AK002280.1 | - | 1 - 2 | 170 | 65 | |
| AK010276.1 | - | 1 - 2 | 134 | 65 | ||
| AK011863.1 | - | 1 - 2 | 179 | 65 | ||
| AK011984.1 | - | 1 - 2 | 191 | 65 | ||
| AK145487.1 | - | 1 - 2 | 191 | 65 | ||
| AK168339.1 | - | 1 - 2 | 169 | 65 | ||
| AK168863.1 | - | 1 - 2 | 188 | 65 | ||
| BC051930.1 | - | 1 - 2 | 131 | 6403128 | 65 | |
| BC060245.1 | - | 1 - 2 | 174 | 6819088 | 65 | |
| BY128235.1 | - | 1 - 2 | 247 | 65 | ||
More Information
Set Membership: |
|

Psmd9
Genomic Map