Assay Details
| Gene Symbol: Anapc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042491.2 | NP_001035956.1 | 4 - 5 | 734 | 76 | |
| NM_001289517.1 | NP_001276446.1 | 4 - 5 | 734 | 76 | ||
| NM_001289518.1 | NP_001276447.1 | 4 - 5 | 734 | 76 | ||
| NM_001289519.1 | NP_001276448.1 | 4 - 5 | 734 | 76 | ||
| NM_001289520.1 | NP_001276449.1 | 4 - 5 | 734 | 76 | ||
| NM_021505.3 | NP_067480.2 | 4 - 5 | 734 | 76 | ||
| GenBank mRNA | AB041593.1 | - | 4 - 5 | 677 | 76 | |
| AK003821.1 | - | 1 - 2 | 127 | 76 | ||
| AK031432.1 | - | 4 - 5 | 659 | 76 | ||
| AK049719.1 | - | 4 - 5 | 688 | 76 | ||
| AK075969.1 | - | 4 - 5 | 673 | 76 | ||
| AK088327.1 | - | 4 - 5 | 697 | 76 | ||
| AK147951.1 | - | 4 - 5 | 673 | 76 | ||
| AK148679.1 | - | 4 - 5 | 689 | 76 | ||
| AK151314.1 | - | 4 - 5 | 680 | 76 | ||
| AK151905.1 | - | 4 - 5 | 680 | 76 | ||
| AK152528.1 | - | 4 - 5 | 650 | 76 | ||
| AK152981.1 | - | 4 - 5 | 680 | 76 | ||
| AK153373.1 | - | 4 - 5 | 680 | 76 | ||
| AK153422.1 | - | 4 - 5 | 680 | 76 | ||
| AK159204.1 | - | 4 - 5 | 680 | 76 | ||
| AK159713.1 | - | 4 - 5 | 673 | 76 | ||
| AK159916.1 | - | 4 - 5 | 673 | 76 | ||
| AK167561.1 | - | 4 - 5 | 673 | 76 | ||
| AK168135.1 | - | 4 - 5 | 658 | 76 | ||
| AK168737.1 | - | 4 - 5 | 674 | 76 | ||
| AK168814.1 | - | 4 - 5 | 673 | 76 | ||
| AK169196.1 | - | 4 - 5 | 697 | 76 | ||
| BC010339.1 | - | 4 - 5 | 650 | 2648173 | 76 | |
| BC046566.1 | - | 4 - 5 | 713 | 2650790 | 76 | |
| BC046804.1 | - | 4 - 5 | 671 | 6404923 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc5
Genomic Map