Assay Details
| Gene Symbol: Kdm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013910.2 | NP_038938.1 | 4 - 5 | 661 | 65 | |
| GenBank mRNA | AK129479.1 | - | 15 - 16 | 2098 | 65 | |
| BC057622.1 | - | 4 - 5 | 656 | 6402440 | 65 | |
| BC065090.1 | - | 15 - 16 | 2297 | 6851421 | 65 | |
| BC068239.1 | - | 12 - 13 | 1851 | 65 | ||
More Information
Set Membership: |
|

Kdm2b
Genomic Map