Assay Details
| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133791.4 | NP_598552.2 | 16 - 17 | 2919 | 113 | |
| GenBank mRNA | AK028956.1 | - | 536 | 113 | ||
| AK032989.1 | - | 12 - 13 | 1985 | 113 | ||
| AK079375.1 | - | 16 - 17 | 2755 | 113 | ||
| AK147648.1 | - | 2913 | 113 | |||
| BC054434.1 | - | 6 - 7 | 1200 | 113 | ||
| BC067050.1 | - | 16 - 17 | 2919 | 30537299 | 113 | |
More Information
Set Membership: |
|

Wwc2
Genomic Map