Assay Details
| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081074.1 | NP_001074543.1 | 10 - 11 | 1626 | 107 | |
| GenBank mRNA | AY566862.1 | - | 10 - 11 | 1623 | 107 | |
| AY566863.1 | - | 10 - 11 | 1599 | 107 | ||
| BC038942.1 | - | 8 - 9 | 1302 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A1cf
Genomic Map