Assay Details
| Gene Symbol: Gper1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029771.3 | NP_084047.2 | 2 - 3 | 384 | 53 | |
| GenBank mRNA | AK018203.1 | - | 2 - 3 | 288 | 53 | |
| AK030375.1 | - | 2 - 3 | 385 | 53 | ||
| BC138598.1 | - | 2 - 3 | 239 | 8861618 | 53 | |
| BC138616.1 | - | 2 - 3 | 239 | 8861636 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018203.1, AK030375.1, BC138598.1, BC138616.1, NM_029771.3 transcripts.
Set Membership: |
|

Gper1
Genomic Map