Assay Details
| Gene Symbol: Taf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081288.1 | NP_001074757.1 | 23 - 24 | 3334 | 64 | |
| GenBank mRNA | AK147903.1 | - | 23 - 24 | 3337 | 64 | |
| BC147638.1 | - | 23 - 24 | 3285 | 9056628 | 64 | |
| BC151079.1 | - | 23 - 24 | 3285 | 9087992 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taf2
Genomic Map