Assay Details
| Gene Symbol: Taf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081288.1 | NP_001074757.1 | 7 - 8 | 1201 | 55 | |
| GenBank mRNA | AK028828.1 | - | 7 - 8 | 948 | 55 | |
| AK147903.1 | - | 7 - 8 | 1203 | 55 | ||
| BC147638.1 | - | 7 - 8 | 1152 | 9056628 | 55 | |
| BC151079.1 | - | 7 - 8 | 1152 | 9087992 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Taf2
Genomic Map