Assay Details
| Gene Symbol: Ppp1r32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005821.1 | - | 4 - 5 | 507 | 72 | |
| AK016325.1 | - | 4 - 5 | 505 | 72 | ||
| BC118523.1 | - | 4 - 5 | 451 | 40105249 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r32
Genomic Map