Assay Details
| Gene Symbol: Klkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008455.2 | NP_032481.2 | 10 - 11 | 1237 | 83 | |
| GenBank mRNA | BC026555.1 | - | 10 - 11 | 1163 | 4972788 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|

Klkb1
Genomic Map