Assay Details
| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033246.2 | NP_001028418.1 | 1 - 2 | 63 | 66 | |
| GenBank mRNA | AK132742.1 | - | 1 - 2 | 64 | 66 | |
| AK161248.1 | - | 1 - 2 | 51 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132742.1, AK161248.1, NM_001033246.2 transcripts.
Set Membership: |
|

Ccdc110
Genomic Map