Assay Details
| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033246.2 | NP_001028418.1 | 3 - 4 | 267 | 71 | |
| GenBank mRNA | AK132742.1 | - | 3 - 4 | 268 | 71 | |
| AK161248.1 | - | 3 - 4 | 255 | 71 | ||
More Information
Set Membership: |
|

Ccdc110
Genomic Map