Assay Details
| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF356627.1 | - | 11 - 12 | 1379 | 133 | |
| AK005546.1 | - | 11 - 12 | 1391 | 133 | ||
| BC019485.1 | - | 11 - 12 | 1368 | 4194672 | 133 | |
More Information
Updated Mapping Information
Set Membership: |
|

F11
Genomic Map