Assay Details
| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF356627.1 | - | 3 - 4 | 305 | 75 | |
| AK005546.1 | - | 3 - 4 | 317 | 75 | ||
| BC019485.1 | - | 3 - 4 | 294 | 4194672 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

F11
Genomic Map