Assay Details
| Gene Symbol: Tmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028953.2 | NP_083229.1 | 7 - 8 | 680 | 60 | |
| GenBank mRNA | AF417579.1 | - | 7 - 8 | 680 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmc1
Genomic Map