Assay Details
| Gene Symbol: Casp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284409.1 | NP_001271338.1 | 5 - 6 | 711 | 79 | |
| NM_009810.3 | NP_033940.1 | 5 - 6 | 706 | 79 | ||
| GenBank mRNA | AK080651.1 | - | 2 - 3 | 221 | 79 | |
| AK146226.1 | - | 5 - 6 | 712 | 79 | ||
| BC038825.1 | - | 5 - 6 | 595 | 4482368 | 79 | |
| D86352.1 | - | 5 - 6 | 525 | 79 | ||
| U19522.1 | - | 5 - 6 | 522 | 79 | ||
| U49929.1 | - | 5 - 6 | 548 | 79 | ||
| U63720.1 | - | 2 - 3 | 314 | 79 | ||
| Y13086.1 | - | 5 - 6 | 552 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp3
Genomic Map