Assay Details
| Gene Symbol: Acsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK145900.1 | - | 1 - 2 | 174 | 85 | |
| AK149406.1 | - | 1 - 2 | 52 | 85 | ||
| AK152772.1 | - | 1 - 2 | 52 | 85 | ||
| AK153050.1 | - | 1 - 2 | 180 | 85 | ||
| AK158388.1 | - | 1 - 2 | 131 | 85 | ||
| AK168078.1 | - | 1 - 2 | 52 | 85 | ||
| BY786934.1 | - | 1 - 2 | 192 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145900.1, AK149406.1, AK152772.1, AK153050.1, AK168078.1 transcripts.
Set Membership: |
|

Acsl1
Genomic Map