Assay Details
| Gene Symbol: Cenpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027973.3 | NP_082249.1 | 6 - 7 | 611 | 60 | |
| GenBank mRNA | AK006479.1 | - | 6 - 7 | 620 | 60 | |
| AK050386.1 | - | 6 - 7 | 612 | 60 | ||
| AK081703.1 | - | 6 - 7 | 599 | 60 | ||
| BC094675.1 | - | 7 - 8 | 731 | 60 | ||
| BC117784.2 | - | 6 - 7 | 612 | 40092112 | 60 | |
| BC117785.2 | - | 6 - 7 | 612 | 40092113 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpu
Genomic Map