Assay Details
| Gene Symbol: Zfat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145888.1 | NP_001139360.1 | 10 - 11 | 3042 | 92 | |
| NM_198644.2 | NP_941046.1 | 10 - 11 | 3021 | 92 | ||
| GenBank mRNA | BC053930.1 | - | 10 - 11 | 3012 | 6389756 | 92 |
| EU221277.1 | - | 10 - 11 | 2872 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfat
Genomic Map