Assay Details
| Gene Symbol: Adgrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174991.3 | NP_778156.2 | 23 - 24 | 4207 | 72 | |
| GenBank mRNA | AK046831.1 | - | 7 - 8 | 725 | 72 | |
| AK147456.1 | - | 23 - 24 | 4192 | 72 | ||
| AK147459.1 | - | 23 - 24 | 4160 | 72 | ||
| AK147494.1 | - | 23 - 24 | 3994 | 72 | ||
| AK147607.1 | - | 23 - 24 | 4211 | 72 | ||
| AK220481.1 | - | 23 - 24 | 3586 | 72 | ||
| AY168408.1 | - | 23 - 24 | 3719 | 72 | ||
| BC036963.1 | - | 5 - 6 | 485 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adgrb1
Genomic Map