Assay Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF229633.1 | - | 2 - 3 | 154 | 83 | |
| AK017548.1 | - | 2 - 3 | 717 | 83 | ||
| AK134989.1 | - | 2 - 3 | 785 | 83 | ||
| BC086631.1 | - | 3 - 4 | 698 | 83 | ||
| BQ769921.1 | - | 2 - 3 | 205 | 83 | ||
| BY073734.1 | - | 2 - 3 | 228 | 83 | ||
| CD549374.1 | - | 2 - 3 | 129 | 83 | ||
More Information
Set Membership: |
|

Tle4
Genomic Map