Assay Details
| Gene Symbol: Tle4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF229633.1 | - | 5 - 6 | 327 | 60 | |
| AK017548.1 | - | 5 - 6 | 889 | 60 | ||
| AK122481.2 | - | 3 - 4 | 186 | 60 | ||
| AK134989.1 | - | 5 - 6 | 958 | 60 | ||
| BC086631.1 | - | 6 - 7 | 871 | 60 | ||
| BQ769921.1 | - | 5 - 6 | 378 | 60 | ||
More Information
Set Membership: |
|

Tle4
Genomic Map