Assay Details
| Gene Symbol: Tmprss6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027902.2 | NP_082178.2 | 13 - 14 | 1794 | 71 | |
| GenBank mRNA | AK004939.2 | - | 13 - 14 | 1641 | 71 | |
| AY240929.1 | - | 13 - 14 | 1771 | 71 | ||
| BC029645.1 | - | 13 - 14 | 1771 | 4195486 | 71 | |
| BC057674.1 | - | 13 - 14 | 1736 | 4162007 | 71 | |
| EU190436.1 | - | 12 - 13 | 1585 | 71 | ||
| EU190437.1 | - | 12 - 13 | 1585 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss6
Genomic Map