Assay Details
| Gene Symbol: Tmprss6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027902.2 | NP_082178.2 | 4 - 5 | 646 | 92 | |
| GenBank mRNA | AK004939.2 | - | 4 - 5 | 493 | 92 | |
| AY240929.1 | - | 4 - 5 | 623 | 92 | ||
| BC029645.1 | - | 4 - 5 | 623 | 4195486 | 92 | |
| BC057674.1 | - | 4 - 5 | 588 | 4162007 | 92 | |
| EU190436.1 | - | 3 - 4 | 437 | 92 | ||
| EU190437.1 | - | 3 - 4 | 437 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss6
Genomic Map