Assay Details
| Gene Symbol: Csf2rb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287389.1 | NP_001274318.1 | 3 - 4 | 506 | 69 | |
| NM_007781.3 | NP_031807.1 | 3 - 4 | 506 | 69 | ||
| GenBank mRNA | AK154650.1 | - | 3 - 4 | 429 | 69 | |
| AK155216.1 | - | 3 - 4 | 69 | |||
| AK155511.1 | - | 3 - 4 | 69 | |||
| AK155626.1 | - | 3 - 4 | 408 | 69 | ||
| AK155675.1 | - | 3 - 4 | 69 | |||
| AK170028.1 | - | 3 - 4 | 69 | |||
| M29855.1 | - | 3 - 4 | 388 | 69 | ||
More Information
Set Membership: |
|

Csf2rb2
Genomic Map