Assay Details
| Gene Symbol: Rfx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166414.1 | NP_001159886.1 | 11 - 12 | 1359 | 66 | |
| NM_011265.3 | NP_035395.2 | 10 - 11 | 1400 | 66 | ||
| GenBank mRNA | AK030008.1 | - | 12 - 13 | 1432 | 66 | |
| AK132267.1 | - | 6 - 7 | 725 | 66 | ||
| AK140928.1 | - | 9 - 10 | 1208 | 66 | ||
| BC017598.1 | - | 10 - 11 | 1374 | 4483833 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rfx3
Genomic Map