Assay Details
| Gene Symbol: Rfx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166414.1 | NP_001159886.1 | 6 - 7 | 705 | 73 | |
| NM_011265.3 | NP_035395.2 | 5 - 6 | 746 | 73 | ||
| GenBank mRNA | AK030008.1 | - | 7 - 8 | 778 | 73 | |
| BC017598.1 | - | 5 - 6 | 720 | 4483833 | 73 | |
| X76090.1 | - | 2 - 3 | 137 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx3
Genomic Map