Assay Details
| Gene Symbol: Mtbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168250.1 | NP_001161722.1 | 4 - 5 | 633 | 115 | |
| NM_134092.3 | NP_598853.3 | 4 - 5 | 633 | 115 | ||
| GenBank mRNA | AK080036.1 | - | 4 - 5 | 447 | 115 | |
| AK088549.1 | - | 4 - 5 | 447 | 115 | ||
| AK133855.1 | - | 4 - 5 | 481 | 115 | ||
| BC052197.1 | - | 4 - 5 | 633 | 6511752 | 115 | |
More Information
Set Membership: |
|

Mtbp
Genomic Map