Assay Details
| Gene Symbol: Mtbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168250.1 | NP_001161722.1 | 9 - 10 | 1165 | 82 | |
| NM_134092.3 | NP_598853.3 | 9 - 10 | 1165 | 82 | ||
| GenBank mRNA | AJ278508.1 | - | 9 - 10 | 1086 | 82 | |
| AK080036.1 | - | 9 - 10 | 979 | 82 | ||
| AK088549.1 | - | 9 - 10 | 979 | 82 | ||
| AK133855.1 | - | 9 - 10 | 1013 | 82 | ||
| BC005500.1 | - | 2 - 3 | 162 | 82 | ||
| BC052197.1 | - | 9 - 10 | 1165 | 6511752 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtbp
Genomic Map