Assay Details
| Gene Symbol: Dscc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183089.2 | NP_898912.2 | 3 - 4 | 551 | 92 | |
| GenBank mRNA | AK052329.1 | - | 1 - 2 | 150 | 92 | |
| AK144502.1 | - | 3 - 4 | 554 | 92 | ||
| BC099382.1 | - | 2 - 3 | 180 | 92 | ||
| BC116833.1 | - | 3 - 4 | 526 | 40126152 | 92 | |
| BC116835.1 | - | 3 - 4 | 526 | 40126154 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dscc1
Genomic Map