Assay Details
| Gene Symbol: Dscc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183089.2 | NP_898912.2 | 4 - 5 | 646 | 97 | |
| GenBank mRNA | AK008137.1 | - | 4 - 5 | 646 | 97 | |
| AK144502.1 | - | 4 - 5 | 649 | 97 | ||
| BC099382.1 | - | 3 - 4 | 275 | 97 | ||
| BC116833.1 | - | 4 - 5 | 621 | 40126152 | 97 | |
| BC116835.1 | - | 4 - 5 | 621 | 40126154 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dscc1
Genomic Map