Assay Details
| Gene Symbol: Dscc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183089.2 | NP_898912.2 | 7 - 8 | 997 | 107 | |
| GenBank mRNA | AK008137.1 | - | 7 - 8 | 997 | 107 | |
| AK011162.1 | - | 7 - 8 | 1014 | 107 | ||
| AK144502.1 | - | 8 - 9 | 1189 | 107 | ||
| BC099382.1 | - | 6 - 7 | 626 | 107 | ||
| BC116833.1 | - | 7 - 8 | 972 | 40126152 | 107 | |
| BC116835.1 | - | 7 - 8 | 972 | 40126154 | 107 | |
| BM940984.1 | - | 1 - 2 | 87 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dscc1
Genomic Map