Assay Details
| Gene Symbol: Lonrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081150.1 | NP_001074619.1 | 6 - 7 | 1676 | 127 | |
| GenBank mRNA | BC094431.1 | - | 3 - 4 | 484 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lonrf1
Genomic Map