Assay Details
| Gene Symbol: Taf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009315.3 | NP_033341.1 | 7 - 8 | 797 | 65 | |
| GenBank mRNA | AK153731.1 | - | 845 | 65 | ||
| AK154867.1 | - | 7 - 8 | 831 | 65 | ||
| BC058583.1 | - | 7 - 8 | 797 | 6514960 | 65 | |
| D49439.1 | - | 6 - 7 | 730 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Taf6
Genomic Map